missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf72 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24522-1-AP-20UL
This item is not returnable.
View return policy
Description
C16orf72 is a protein coding gene.Specifications
C16orf72 | |
Polyclonal | |
Unconjugated | |
C16orf72 | |
C16orf72, PRO0149, UPF0472 protein C16orf72 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29035 | |
-20°C | |
Liquid |
Western Blot | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14CZ0 | |
C16orf72 | |
C16orf72 Fusion Protein Ag21585 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction