missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf72 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1081.84 DKK - 3058.99 DKK
Specifications
Antigen | C16orf72 |
---|---|
Concentration | 0.55 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16862995
|
Proteintech
24522-1-AP-20UL |
20 μL |
1081.84 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16852995
|
Proteintech
24522-1-AP-150UL |
150 μL |
3058.99 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C16orf72 is a protein coding gene.Specifications
C16orf72 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q14CZ0 | |
29035 | |
C16orf72 Fusion Protein Ag21585 | |
Primary | |
-20°C | |
C16orf72 |
0.55 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C16orf72, PRO0149, UPF0472 protein C16orf72 | |
C16orf72 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title