Learn More
Tenak™ Comfort Rack for Microtiter Plates for Upright Freezers, 85mm Shelf Height
Providing a complete range of racks for microtiter plates in chest freezers. Tenak™ Comfort Racks for Microtiter Plates in Upright Freezers are stackable.
Brand: Tenak™ TE35442
1234.79 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Racks can either be used as one plate per shelf or the plates can be stacked
- With an alert preventing the shelves from sliding out
- External footprint is 135 x 92mm
Specifications
Microtiter Plate Rack | |
Plates (12) | |
Microtiter Plates, Upright Freezers | |
360 x 135 x 259 mm | |
259 mm | |
360 mm |
3 x 4 | |
Yes | |
1 Pack | |
135 mm | |
85 mm |