Learn More
Tenak™ Comfort Rack for Microtiter Plates for Upright Freezers, 85mm Shelf Height
Providing a complete range of racks for microtiter plates in chest freezers. Tenak™ Comfort Racks for Microtiter Plates in Upright Freezers are stackable.
1060.00 DKK - 1970.00 DKK
Specifications
Product Type | Microtiter Plate Rack |
---|---|
Stackable | Yes |
For Use With (Equipment) | Microtiter Plates, Upright Freezers |
No. per Pack | 1 Pack |
Width (Metric) | 135 mm |
Product Code | Brand | Holds | Dimensions (L x W x H) | Array | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Dimensions (L x W x H) | Array | Price | Quantity & Availability | |||
12417070
|
Tenak™
TE35510 |
Plates (10) | 450 x 135 x 173 mm | 2 x 5 |
1130.00 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||
12427070
|
Tenak™
TE35512 |
Plates (15) | 450 x 135 x 259 mm | 3 x 5 |
1345.00 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||
12427090
|
Tenak™
TE35560 |
Plates (12) | 540 x 135 x 173 mm | 2 x 6 |
1270.00 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||
12427110
|
Tenak™
TE35610 |
Plates (14) | 630 x 135 x 173 mm | 2 x 7 |
1410.00 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||
12437070
|
Tenak™
TE35514 |
Plates (20) | 450 x 135 x 346 mm | 4 x 5 |
1550.00 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||
12437090
|
Tenak™
TE35562 |
Plates (18) | 540 x 135 x 259 mm | 3 x 6 |
1550.00 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||
12437110
|
Tenak™
TE35612 |
Plates (21) | 630 x 135 x 259 mm | 3 x 7 |
1690.00 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||
12447090
|
Tenak™
TE35564 |
Plates (24) | 540 x 135 x 346 mm | 4 x 6 |
1830.00 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||
12447110
|
Tenak™
TE35614 |
Plates (28) | 630 x 135 x 346 mm | 4 x 7 |
1970.00 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||
12477040
|
Tenak™
TE35440 |
Plates (8) | 360 x 135 x 173 mm | 2 x 4 |
1060.00 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||
12487040
|
Tenak™
TE35442 |
Plates (12) | 360 x 135 x 259 mm | 3 x 4 |
1270.00 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
|||
Description
- Racks can either be used as one plate per shelf or the plates can be stacked
- With an alert preventing the shelves from sliding out
- External footprint is 135 x 92mm
Specifications
Microtiter Plate Rack | |
Microtiter Plates, Upright Freezers | |
135 mm |
Yes | |
1 Pack | |
85 mm |