missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated OLIG2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66513-20UL
This item is not returnable.
View return policy
Description
The OLIG2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human OLIG2. This antibody recognizes human, mouse, rat antigen. The OLIG2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.Specifications
OLIG2 | |
Monoclonal | |
1000 μg/mL | |
BHLHB1; OLIGO2; PRKCBP2; RACK17; bHLHe19 | |
Recombinant Protein | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
35-39 kDa | |
Liquid |
Immunofluorescence | |
1E8C5 | |
CoraLite Plus 488 | |
Mouse | |
Protein A purification | |
RUO | |
10215 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Conjugated Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction