missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated OLIG2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
968.42 DKK - 2619.89 DKK
Specifications
Antigen | OLIG2 |
---|---|
Clone | 1E8C5 |
Concentration | 1000 μg/mL |
Applications | Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30235155
|
Proteintech
CL488-66513-100UL |
100 μL |
2619.89 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30237460
|
Proteintech
CL488-66513-20UL |
20 μL |
968.42 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The OLIG2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human OLIG2. This antibody recognizes human, mouse, rat antigen. The OLIG2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.Specifications
OLIG2 | |
1000 μg/mL | |
Monoclonal | |
Liquid | |
RUO | |
10215 | |
Recombinant Protein | |
Primary | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
1E8C5 | |
Immunofluorescence | |
CoraLite Plus 488 | |
Mouse | |
Human, Mouse, Rat | |
BHLHB1; OLIGO2; PRKCBP2; RACK17; bHLHe19 | |
IgG2a | |
Protein A purification | |
Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title