missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Syntaxin 6 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60059-20UL
This item is not returnable.
View return policy
Description
The Syntaxin 6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Syntaxin 6. This antibody recognizes human, mouse, rat antigen. The Syntaxin 6 antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
Syntaxin 6 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Protein G purification | |
RUO | |
10228 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
Conjugated Antibody | |
IgG1 |
Immunofluorescence | |
2B6G4 | |
CoraLite Plus 488 | |
Recombinant Protein | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
29 kDa | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction