Learn More
SP Bel-Art™ Durac™ Calibrated Electronic Stainless Steel Stem Thermometer
Suitable for field studies, wet lab applications, wash down areas, food testing and tanks
Brand: SP Bel-Art™ B60900-3600
Description
- Reads temperatures in liquids, semi-solids and gases
- Stainless-steel probe easily reaches into cuvettes, test tubes, flasks, beakers, etc.
- Includes accredited calibration documentation that meets the requirements of ISO 17025
- Individually serialized
- CE marked
Certifications
Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
Specifications
0, 100 °C | |
Reading temperatures in liquids, semi-solids and gases | |
3 in. | |
76 mm | |
IP67 | |
1.2 in. | |
7.1 in. | |
-58 to 392 °F | |
CR2032 | |
30 mm |
CE marked | |
Accredited calibration documentation, Multi-language instructions | |
180 mm | |
Stainless Steel | |
0.1 °C/F | |
Digital thermometer | |
Waterproof | |
-50 to 200 °C | |
±0.5°C from 10/100°C, ±1.5°C |
Electronic stem thermometers are not suitable for solids such as frozen items