Learn More
SP Bel-Art™ Durac™ Calibrated Electronic Stainless Steel Stem Thermometer
Suitable for field studies, wet lab applications, wash down areas, food testing and tanks
468.00 DKK - 636.00 DKK
Specifications
Calibration Points | 0, 100 °C |
---|---|
Certifications/Compliance | CE marked |
For Use With (Application) | Reading temperatures in liquids, semi-solids and gases |
Includes | Accredited calibration documentation, Multi-language instructions |
Material | Stainless Steel |
Product Code | Brand | Description | Length (English) Probe | Model | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Length (English) Probe | Model | Price | Quantity & Availability | |||
17294883
|
SP Bel-Art™
B60900-3600 |
Waterproof | 3 in. | IP67 | N/A | ||||
17204893
|
SP Bel-Art™
B60900-3700 |
Water resistant | 4.8 in. | IP66 |
636.00 DKK
Each |
Estimated Shipment: 15-08-2025
Log in to see stock. |
|||
Description
- Reads temperatures in liquids, semi-solids and gases
- Stainless-steel probe easily reaches into cuvettes, test tubes, flasks, beakers, etc.
- Includes accredited calibration documentation that meets the requirements of ISO 17025
- Individually serialized
- CE marked
Certifications
Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
Specifications
0, 100 °C | |
Reading temperatures in liquids, semi-solids and gases | |
Stainless Steel | |
Digital thermometer | |
-50 to 200 °C |
CE marked | |
Accredited calibration documentation, Multi-language instructions | |
0.1 °C/F | |
-58 to 392 °F |
Electronic stem thermometers are not suitable for solids such as frozen items