Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 25 mm Shelf Height
Eco AluRack for chest freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE15142
981.71 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12418180
  • 1010.00 DKK / Each
Only null left
Estimated Shipment: 06-10-2025
Description
  • For use in chest freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications
Specifications
Aluminum
Chest Frseezers
135 x 92 x 620 mm
Freezer Rack
Microtiterplates (23)
25 mm