Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 25 mm Shelf Height
Eco AluRack for chest freezers is designed to hold microtiterplates.
651.00 DKK - 1130.00 DKK
Specifications
Material | Aluminum |
---|---|
Type | Freezer Rack |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 25 mm |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12408180
|
Tenak™
TE15140 |
Microtiterplates (21) |
954.00 DKK
Each |
Estimated Shipment: 06-10-2025
Log in to see stock. |
|||||
12418180
|
Tenak™
TE15142 |
Microtiterplates (23) |
1010.00 DKK
Each |
Estimated Shipment: 06-10-2025
Log in to see stock. |
|||||
12438180
|
Tenak™
TE15146 |
Microtiterplates (27) |
1130.00 DKK
Each |
Estimated Shipment: 06-10-2025
Log in to see stock. |
|||||
12458170
|
Tenak™
TE15130 |
Microtiterplates (11) |
651.00 DKK
Each |
Estimated Shipment: 06-10-2025
Log in to see stock. |
|||||
12468170
|
Tenak™
TE15132 |
Microtiterplates (13) |
708.00 DKK
Each |
Estimated Shipment: 06-10-2025
Log in to see stock. |
|||||
12488170
|
Tenak™
TE15136 |
Microtiterplates (17) |
831.00 DKK
Each |
Estimated Shipment: 06-10-2025
Log in to see stock. |
|||||
12498170
|
Tenak™
TE15138 |
Microtiterplates (19) |
892.00 DKK
Each |
Estimated Shipment: 06-10-2025
Log in to see stock. |
|||||
Description
- For use in chest freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
Aluminum | |
Freezer Rack | |
Chest Frseezers | |
25 mm |