Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 25 mm Shelf Height
Eco AluRack for chest freezers is designed to hold microtiterplates.
Brand: Tenak™ TE15136
807.20 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use in chest freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
Aluminum | |
Chest Frseezers | |
135 x 92 x 461 mm |
Freezer Rack | |
Microtiterplates (17) | |
25 mm |