Learn More
Macherey-Nagel™ N9 Polypropylene Blue Screw Caps
Seal vials with screw caps with a secure septum to receive needles. The Macherey-NagelTM N9 pollypropylene blue screw caps come with a choice of septa.
Brand: Macherey-Nagel™ 702288.1
220.10 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Screw closures N 9
- Large range of screw closures with different septum materials and cap colors
- Bonded screw caps (septa firmly connected with the cap; cannot be removed) – advantage: thick (blunt) HPLC needles cannot push the septum into the vial
- Polymagnetic screw closure 702166 for CTC GC PAL
Specifications
Blue | |
Yes | |
1 mm |
Polypropylene, PTFE, Silicone | |
White Silicone/Blue PTFE | |
N9 |