Learn More
Macherey-Nagel™ N9 Polypropylene Blue Screw Caps
Seal vials with screw caps with a secure septum to receive needles. The Macherey-NagelTM N9 pollypropylene blue screw caps come with a choice of septa.
131.00 DKK - 239.00 DKK
Specifications
Color Blue
Thickness (Metric) 1 mm
Closure Size N9
View More Specs
Products 3
Product Code Brand Septum Price Quantity & Availability  
Product Code Brand Septum Price Quantity & Availability  
10411708
View Documents
Macherey-Nagel™
702732
Red Rubber/Clear FEP
131.00 DKK
Pack of 100
View alternative products
Save Up to 
»
Only null left
In Stock
Add to basket
10282660
View Documents
Macherey-Nagel™
702287.1
White Silicone/Red PTFE
196.00 DKK
Pack of 100
View alternative products
Save Up to 
»
Only null left
In Stock
Add to basket
10327563
View Documents
Macherey-Nagel™
702288.1
White Silicone/Blue PTFE
239.00 DKK
Pack of 100
View alternative products
Save Up to 
»
Only null left
In Stock
Add to basket
Description
Description
  • Screw closures N 9
  • Large range of screw closures with different septum materials and cap colors
  • Bonded screw caps (septa firmly connected with the cap; cannot be removed) – advantage: thick (blunt) HPLC needles cannot push the septum into the vial
  • Polymagnetic screw closure 702166 for CTC GC PAL
Specifications
Specifications
Blue
1 mm
N9