missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC36A1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24775-1-AP-150UL
This item is not returnable.
View return policy
Description
The SLC36A1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC36A1. This antibody recognizes human antigen. The SLC36A1 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
SLC36A1 | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Antigen affinity purification | |
RUO | |
206358 | |
Store at -20°C. Stable for one year after shipment. | |
Polyclonal Antibody | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
1000 μg/mL | |
LYAAT1; PAT1; TRAMD3 | |
Recombinant Protein | |
150 μL | |
Primary | |
Human | |
43-53 kDa | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction